• USA Home
  • Z559032 - Sigma-Aldrich® anti-static disposable unisex lab coat
EMAIL THIS PAGE TO A FRIEND
Z559032 Sigma-Aldrich
Sigma-Aldrich® anti-static disposable unisex lab coat
light blue, size 3XL
  •  NACRES NB.35
Purchase
Properties
Related Categories Anti-Static, Anti-Static Disposable Laboratory Coats, Disposable Lab Coats, Lab Coats, Labware,
size   3XL
availability   not available in EU
color   light blue
Description
General description
DL4260 Disposable Lab Coats. These tough, lightweight, full-length lab coats are splash/spray resistant and offer superior comfort/breathability, and are anti-static treated.
Details:
• Material: SMS
• Weight: 30 grams
• Collar: knit
• Pockets: 3
• Cuffs, Heavy Duty Snaps, Latex Free: yes
Legal Information
Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?