• USA Home
  • Z559059 - Sigma-Aldrich® anti-static disposable unisex lab coat
EMAIL THIS PAGE TO A FRIEND
Z559059 Sigma-Aldrich
Sigma-Aldrich® anti-static disposable unisex lab coat
white, size 5XL
  •  NACRES NB.35
Purchase
Properties
Related Categories Anti-Static, Anti-Static Disposable Laboratory Coats, Disposable Lab Coats, Lab Coats, Labware,
size   5XL
availability   not available in EU
color   white
Description
General description
DL4260 Disposable Lab Coats. These tough, lightweight, full-length lab coats are splash/spray resistant and offer superior comfort/breathability, and are anti-static treated.
Details:
• Material: SMS
• Weight: 30 grams
• Collar: knit
• Pockets: 3
• Cuffs, Heavy Duty Snaps, Latex Free: yes
Legal Information
Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?