• USA Home
  • Z565733 - Sleeve stopper septa

Z565733 Aldrich

Sleeve stopper septa

bottom I.D. × O.D. 6.5 mm × 13.3 mm, white



Related Categories Labware, Septa, Sleeve Stopper Septa More...
material   natural rubber
feature   Use with 13 mm I.D. tubes
bottom I.D. × O.D.   6.5 mm × 13.3 mm
color   white


General description

Sleeve stopper septa are manufactured from pure natural rubber for excellent resealability when used for cannulation techniques or from FKM rubber for high chemical resistance. Natural rubber septa are available in red and white colors and a soft rubber formulation that contains no fillers or coloring agents for super tactility and easy manipulation.

Storage Note for Natural Rubber Septa:
Store septa in a closed bag out of sunlight to retard blooming, which is the formation of a whitish film on the surface of the rubber. Blooming is a natural process and does not affect performance. Remove visible bloom by washing or wiping off before use.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.


Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?