• USA Home
  • Z567728 - High pressure adapter for Q-Tube
EMAIL THIS PAGE TO A FRIEND
Z567728 Aldrich
High pressure adapter for Q-Tube
for 12 mL Q-Tube
Synonym: humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
Purchase
Properties
Related Categories Labware, Pressure Vessels, Q-Tube Pressure Tube Reactors More...
feature   pressure 200 psi max.
mfr. no.   Q Labtech
  Q Labtech
Description
Legal Information
Q-Tube is a trademark of Q Labtech LLC
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Frequently Asked Questions
Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
How do I get lot-specific information or a Certificate of Analysis?
The lot specific COA document can be found by entering the lot number above under the "Documents" section.
How do I find price and availability?
There are several ways to find pricing and availability for our products.Once you log onto our website, you will find the price and availability displayed on the product detail page.You can contact any of our Customer Sales and Service offices to receive a quote. USA customers: 1-800-325-3010 orview local office numbers.
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
What are the benefits of High pressure adapters for Q-Tube?
The Q-TubeTM reactors are affordable alternatives to a microwave synthesizer. Unlike the microwave system, which can experience hot-spots that can lead to charring of product, the Q-TubeTM system provides more uniform heat distribution.
Can High pressure adapters for Q-Tube be heated with an oil bath?
Yes. However, the Q-BlockTM heating block offers the advantage of being equipped with a safety shield, lattice clamps and hose barb for nitrogen stream to cool down the block.
What is the maximum liquid fill level for the High pressure adapter for Q-Tube glass tube?
We would recommend keeping the level of the liquid just below half full. The 35mL system has a maximum fill line printed on the tube.
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.
Show more questions
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?