• USA Home
  • Z647454 - Pyrex® measuring cylinder with spout, heavy duty, squat form
EMAIL THIS PAGE TO A FRIEND
Z647454 Aldrich
Pyrex® measuring cylinder with spout, heavy duty, squat form
100 mL volume, accuracy: 1.0 mL
Purchase
Properties
feature   graduations 2.0 mL
measuring range   100 mL volume, accuracy: 1.0 mL
H   147 mm
Description
General description
Heavy duty construction includes: thicker walls, heavily beaded rim and spout, and robust hexagonal foot.
• Squat height is more stable
• More convenient to use in confined spaces
• Graduations in blue enamel
Legal Information
PYREX is a registered trademark of Corning, Inc.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?