• USA Home
  • Z655090 - Carbolite® tube furnace end seals
EMAIL THIS PAGE TO A FRIEND
Z655090 Aldrich
Carbolite® tube furnace end seals
omit gas inlet (from other end), diam. ~80 mm
Purchase
Properties
Related Categories Carbolite, Heating, Labware, Manufacturer Browser, Ovens More...
mfr. no.   Carbolite 41-38-2
diam.   ~80 mm
Description
General description
The vacuum flange attaches to the stainless steel end plate of the gas-tight end seals, Z655058. Gas tight end seals must be ordered separately and in conjunction with this product for them to work properly. Also required but not supplied Z655082.
Legal Information
Carbolite is a registered trademark of Carbolite Limited
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products
recommended
Product #
Description
Add to Cart
Z655058 Carbolite® tube furnace end seals, gas tight end seal, diam. ~32 mm
Z655082 Carbolite® tube furnace end seals, 1x NW16 flange, diam. ~32 mm
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?