• USA Home
  • Z655503 - Hanna combination ISE electrodes
EMAIL THIS PAGE TO A FRIEND
Z655503 Aldrich
Hanna combination ISE electrodes
Cupric, Solid-state
Purchase
Properties
Related Categories Hanna, Hanna ISE electrodes, Hanna pH Electrodes, Hanna pH Meters and Accessories, Labware,
material   polyetherimide body
mfr. no.   Hanna HI 4108
measuring range   1-1x10 M ISE
  2-12.5 pH
parameter   0-80 °C temp. range
electrode immersion depth × O.D.   120 mm × 12 mm
Description
Application
Used as an indicator for titrations using chelates.
General description
These include a sensor and reference electrode in a single electrode body. Hanna combination ion selective electrodes provide selectivity and include superior double junction reference into an electrode body. Combination solid-state electrodes have a built-in solid-state sensor and quick refill reference electrode.
Hanna′s liquid membrane and fluoride combination electrodes have replaceable module construction and the Hanna double junction refence stability.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?