• USA Home
  • Z671746 - Stuart digital rocker & roller mixers
EMAIL THIS PAGE TO A FRIEND
Z671746 Aldrich
Stuart digital rocker & roller mixers
Stuart SRT6D, AC/DC input 230 V AC (supplied with both UK and EU plugs)
Purchase
Properties
Related Categories Labware, Rockers and Rollers More...
feature   CE compliant
mfr. no.   Stuart SRT6D
parameter   10 kg load
  5-60 rpm speed
Description
General description
These roller mixers provide a gentle, but highly efficient rocking and rolling action.
There are two models available with either 6 or 9 rollers.
The 6 roller design has a small space saving footprint, while the 9 roller unit has a higher capacity. Both units come with digitally controlled speeds between 5 and 60 rpm (includes programmable timer).
Designed for easy cleaning with plastic rollers and drip trays made from stainless steel to hold accidental spillages.
Recommended for mixing blood samples, viscous substances and liquid-solid suspensions where minimum aeration is required. Units can be used in incubators up to 60°C and humidity up to 80%, or in cold rooms down to 4°C.
• Rocking and rolling action for complete mixing
• Designed for continuous quiet operation
• Can be used in cold rooms or in incubators
• Ideal for small vials or bottles (max bottle diam. 135 mm)
Legal Information
Stuart is a trademark of Bibby Sterlin Ltd.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?