EMAIL THIS PAGE TO A FRIEND
Z672106

DYNAMO flashlight

size 50.8 mm × 146 mm, CE compliant

Purchase

Properties

Related Categories Labware, Timers More...
feature   CE compliant
  CSA approved
size   50.8 mm × 146 mm
weight   226.8 g

Description

General description

• Flashlight with 3 white LED ultra bright beams
• 1 minute of cranking offers 30 minutes of continuous bright light for 3 LEDs
• Hand crank generator recharges the lifetime internal battery - To recharge, crank handle
• In proper use - battery can be recharged 500 times - use more than 10 years with once week operation
• Maximum brightness: 60,000 mcd
• Individually packaged

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?