• USA Home
  • Z677329 - Dermatril LR nitrile gloves
EMAIL THIS PAGE TO A FRIEND
Z677329 Aldrich
Dermatril LR nitrile gloves
size M
  •  NACRES NB.35
Purchase
Properties
Related Categories Anti-Static Disposable Nitrile Gloves, Dermatril LR nitrile gloves, Disposable Gloves, Disposable Nitrile Gloves, Gloves,
material   blue gloves
  blue nitrile
size   M
sterility   non-sterile
  non-sterile
feature   CE compliant
  latex free
  powder-free
packaging   pack of 50 ea (boxes of 50)
mfr. no.   KCL 742
  KCL, 742 - 08
L × thickness   270 mm × 0.11 mm
Description
General description
Dermatril LR is an extended cuff, blue nitrile, powder-free glove providing a high level of protection against chemical and bacteriological contamination. The Dermatril LR disposable glove also displays very good physical durability in comparison to conventional disposable gloves.
All Dermatril LR disposable gloves are INDIVIDUALLY marked with the appropriate standards mark applied on the inside using a food-friendly dye.
• Cleanroom class 100
• Anti-static
• Free from allergenic substances such as thiurames, benzothiazoles and thiourea
• Extended cuff
• Roughened fingertips for better grip
• Rolled cuff edge for easier donning
• CE compliant
• Meet standards: EN 374 (cat. III), EN 388 (cat. II), EN 455, EN 61340-5-1, ASTM 1671:2001, US-FED-ST 209E
Legal Information
Dermatril is a trademark of KCL GmbH
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Content
Coronavirus COVID-19 (SARS-CoV-2) Detection, Characterization & Vaccine Production
As a global life science tools and equipment supplier, we are here to help in the response to COVID-19. From supplying raw materials and research products for detection and characterization to produc...
Keywords: PAGE
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?