• USA Home
  • Z679224 - Microgard 2500 lab coat
EMAIL THIS PAGE TO A FRIEND
Z679224 Aldrich
Microgard 2500 lab coat
size M
  •  NACRES NB.35
Purchase
Properties
Related Categories Disposable Lab Coats, Lab Coats, Lab Coats for Biological and Radioactive Protection, Labware, Safety Clothing,
size   M
feature   CE compliant
color   white
Description
General description
Microgard 2500 is designed to protect from saturating liquid spray and particulates, the fabric′s physical strength and flexibility ensure you are protected yet comfortable.
Features:
• Collar
• Stud front fastening
• Left breast pocket
• Lower right pocket
• Bound seams
• Designed as a single use/disposable product
Legal Information
Microgard is a trademark of Microgard Limited
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Content
Coronavirus COVID-19 (SARS-CoV-2) Detection, Characterization & Vaccine Production
As a global life science tools and equipment supplier, we are here to help in the response to COVID-19. From supplying raw materials and research products for detection and characterization to produc...
Keywords: PAGE
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?