• USA Home
  • Z681865 - CellCrown inserts

EMAIL THIS PAGE TO A FRIEND
Z681865 Aldrich

CellCrown inserts

12 well plate inserts with 1.2 μm nylon filter, sterile

Synonym: 3D Cell Culture, 3D scaffolds, 3D, cell scaffolds

Purchase

Properties

Related Categories 3D Cell Culture Products, 3D Scaffolds, Cell Culture Supplies, Labware, Scaffdex Culture Products More...
sterility   sterile
feature   lid
mfr. no.   Scaffdex, C12002F
size   12 wells
color   none
matrix active group   non-treated surface treatment

Description

General description

CellCrown 12 is available as 5 models; non-sterile and sterile empty inserts and with preattached, white nylon membrane with pore size options of 1.2 and 5 μm and transparent PET filter with pore size of 8 μm. Nylon filter is hydrophilic and binds proteins. PET is specifically developed for cell culturing and it is the right material choice for the filter when smooth cell attachment on the membrane is wanted.

CellCrown is a disposable insert, which immobilizes a sample in its place on a 6, 12, 24, 48, and 96-well plate. Fibers, foils, membranes, textiles, tissue and block like samples can be fixed with CellCrown in a firm, adjusted position in the well. The sample can be left in the well during the change of media by means of a pipette between the wall of the well and the CellCrown. The lid of the well plate can be closed while CellCrown is used. The insert can be removed from the well plate and unloaded after cell culturing without damaging the sample. CellCrown is made of medical grade polycarbonate and the product range includes sterile products ready to use as well as non-sterile products to be sterilized by the user. Sterilization can be by gamma or e-beam radiation, autoclaving, ethylene oxide gas and plasma sterilization.

Sterilization Conditions:
Temperature.......121 °C / 250 °F
Time..................15 min.
Drying time.........15 min.

Legal Information

CellCrown is a trademark of Scaffdex Oy

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?