• USA Home
  • Z682047 - Scaffdex CellVessel insert scaffolds

EMAIL THIS PAGE TO A FRIEND
Z682047 Aldrich

Scaffdex CellVessel insert scaffolds

multifilament copolymer fiber scaffold, diam. 2.9 mm, sterile

Synonym: 3D Cell Culture, 3D scaffolds, 3D, cell scaffolds

Purchase

Properties

Related Categories 3D Cell Culture Products, 3D Scaffolds, Cell Culture Supplies, Labware, Scaffdex Culture Products More...
sterility   sterile
mfr. no.   Scaffdex, V00003
diam.   2.9 mm
fiber L   100 mm
color   white
matrix active group   Non-treated surface treatment

Description

General description

CellVessel is made of bioabsorbable multifilament (8-filaments) poly-96L/4D-lactide copolymer fiber. Scaffdex′s braided scaffold is made for research purposes, only and it should not be used in any kind of clinical settings. Poly-96L/4D-lactide copolymer has been tested with various cell types and it works especially well with fibroblasts.

CellVessel scaffolds are provided as gamma sterilized and with three widths. The shape of the cross-section of CellVessel varies a little bit from complete round to oval, depending on the size. If further sterilization is needed before cell culturing, ethylene oxide or ethanol rinse to avoid further decreasing of inherent viscosity and strenth properties is suggested.

Note: Should CellVessels be cut into wanted lengths by the use, a light heat treatment of the ends of the two bigger sizes should be done to avoid the unloading of the structure during testing.

Legal Information

CellVessel is a trademark of Scaffdex Oy

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?