• USA Home
  • Z682349 - Ace rotary evaporator condenser complete "A" assembly
EMAIL THIS PAGE TO A FRIEND
Z682349
Ace rotary evaporator condenser complete "A" assembly
glass, fits Büchi® models R-200/R-205 and R-114/R-144, includes 1 L receiving flask and clamp
  •  NACRES NB.43
Purchase
Properties
Related Categories Aldrich Evaporator Glassware, Condenser "A" Assemblies, Condenser "A" Assembly and Parts, Evaporators, Labware More...
material   glass
mfr. no.   Büchi equivalent, 40630
Description
General description
Includes 1 L receiving flask and clamp.
The complete "A" evaporator condenser assembly includes the main glass condenser body with inner coil for angled installation, GL threaded caps and hose barbs. Bottom joint is 35/20. It is designed to fit Büchi® evaporator models 200/205 and series 114-144.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?