• USA Home
  • Z682489 - Ace vapor duct tube, for condenser "A" assembly
EMAIL THIS PAGE TO A FRIEND
Z682489
Ace vapor duct tube, for condenser "A" assembly
Joint: ST/NS 29/42, L 160 mm, fits Büchi® models R-114/R-144
  •  NACRES NB.43
Purchase
Properties
Related Categories Aldrich Evaporator Glassware, Evaporators, Labware, Vapor Duct Tubes
Joint   Joint: ST/NS29/42
mfr. no.   Büchi equivalent, 32338
L   160 mm
Description
General description
The vapor duct tube accessories are for Büchi® condenser "A" assemblies, and will fit either Büchi evaporator models R-210/R-215 or R-114/R-144.
Combi-clips: not included.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?