• USA Home
  • 850344P - 14:0-04:0(biphenyl) PC (TBBPC)

EMAIL THIS PAGE TO A FRIEND
850344P Avanti

14:0-04:0(biphenyl) PC (TBBPC)

1-myristoyl-2-[4-(4-biphenyl)butanoyl]-sn-glycero-3-phosphocholine, powder

Synonym: TBBPC

  • CAS Number 865352-99-4

  • Empirical Formula (Hill Notation) C38H60NO8P

  • Molecular Weight 689.86

  •  NACRES NA.25

Purchase

Properties

Related Categories Avanti PC & LPC, Avanti Phospholipids, Avanti Polar Lipids Products, Biochemicals and Reagents, Lipids More...
assay   >99% (TLC)
form   powder
packaging   pkg of 1 × 10 mg (850344P-10mg)
mfr. no.   Avanti Polar Lipids, 850344P
shipped in   dry ice
storage temp.   −20°C
SMILES string   [O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCC1=CC=C(C2=CC=CC=C2)C=C1)=O)COC(CCCCCCCCCCCCC)=O)=O

Description

Packaging

5 mL Clear Glass Sealed Ampule (850344P-10mg)

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Storage & Handling of Lipids - Avanti® Polar Lipids

The following statements pertain specifically to Avanti’s lipid products for research and should not necessarily be applied to cGMP lipids intended for pharmaceutical or diagnostic applications.
Keywords: Diagnostic, Oxidations, Pharmaceutical, Solvents

Protocols

Determination of Phospholipid Oxidation by UV/VIS Spectroscopy - Avanti® Polar Lipids

The percent oxidation of a phospholipid can be determined by using Beer’s Law to calculate the peroxidized phospholipid’s concentration from its absorbance at 234 nm.
Keywords: Extinction coefficient, Oxidations

General Lipid/BSA Solubilization Protocol For Cell Delivery

Water Solutions: Solubilizing some lipids is difficult using organic solvents. Typically, lipids are suspended in methanol to aliquot, the methanol is then dried and the residue dispersed using fatty...
Keywords: Evaporation, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?