• USA Home
  • A-011 - (±)-Amphetamine solution

EMAIL THIS PAGE TO A FRIEND
A-011 Supelco

(±)-Amphetamine solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym: (±)-Amphetamine solution

Purchase

Properties

Related Categories Additional Standards, Amphetamines, Amphetamines / Stimulants Standards, Analytical Standards, Analytical/Chromatography,
grade   certified reference material
InChI Key   KWTSXDURSIMDCE-UHFFFAOYSA-N
feature   SNAP-N-SPIKE®. SNAP-N-SHOOT®
packaging   ampule of 1 mL
mfr. no.   Cerilliant®
drug control   Narcotic Licence Schedule A (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)
concentration   100 μg/mL in methanol
Featured Industry   Pharmaceutical (small molecule)
format   single component solution
storage temp.   −20°C

Description

General description

Amphetamine, a stimulant of the phenethylamine class, is found in ADHD-treatment drugs such as Adderall®, Vyvanse®, and ProCentra®. Because of its stimulant effect, amphetamine is used recreationally as a performance-enhancer and illicit drug. This Certified Spiking Solution® is applicable for use in urine drug testing, sports testing, clinical toxicology, or forensic analysis applications by LC-MS/MS or GC/MS.

Legal Information

Adderall is a registered trademark of Richwood Pharmaceutical Company, Inc.

CERILLIANT is a registered trademark of Cerilliant Corporation

CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation

ProCentra is a registered trademark of Outlook Pharmaceuticals, Inc.

Snap-N-Shoot is a registered trademark of Cerilliant Corporation

Snap-N-Spike is a registered trademark of Cerilliant Corporation

Vyvanse is a registered trademark of Shire, LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Amphetamines-weight loss drugs

Sigma-Aldrich's Cerilliant brand provides the widest selection of Certified Spiking Solutions® for amphetamines and weight loss drugs including native and stable-labeled analogs of amphetamine, metha...
Keywords: Ion-exchange chromatography

Medication Monitoring Testing

Prescription monitoring requires accurate reference standards to ensure consistent and reliable results for drug confirmation and monitoring compliance.
Keywords: Anti-depressants, Anticonvulsants, Cardiovascular, Clinical, Controlled substances, Diabetes, Forensic, Gas chromatography, High performance liquid chromatography, Ion-exchange chromatography, Mass spectrometry, Metabolites, Pharmaceutical, Schizophrenia

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

A-016 (±)-Amphetamine-d11 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
A-050 Amine Mixture-6 solution, 250 μg/mL each component in methanol, ampule of 1 mL, certified reference material, Cerilliant®
A-002 (±)-Amphetamine-d5 (deuterium label on ring) solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
A-005 (±)-Amphetamine-d5 (deuterium label on side chain) solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
A-007 (±)-Amphetamine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
A-008 S(+)-Amphetamine (dextro-Amphetamine) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
A-013 (±)-Amphetamine-d5 (deuterium label on side chain) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
A-017 (±)-Amphetamine-d8 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
A-018 (±)-Amphetamine-d8 solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
A-019 (±)-Amphetamine-d11 solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
A-038 (±)-Amphetamine-d10 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
A-044 (±)-Amphetamine-d6 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
A-045 (±)-Amphetamine-d6 solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
A-049 R(-)-Amphetamine (levo-Amphetamine), 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
E-018 (±)-N-Ethylamphetamine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
L-026 Lisdexamfetamine dimesylate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
L-028 Lisdexamfetamine-D4 dimesylate solution, 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
P-050 PMA hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?