• USA Home
  • A-131 - Androsterone-D4 glucuronide solution

EMAIL THIS PAGE TO A FRIEND
A-131 Supelco

Androsterone-D4 glucuronide solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

  • Empirical Formula (Hill Notation) C25D4H34O8

  • Molecular Weight 470.59

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Pharmaceuticals, Illicit Drugs & Alcohol, Steroids / Hormones
Quality Level   300
grade   certified reference material
feature   Snap-N-Spike®/Snap-N-Shoot®
packaging   ampule of 1 mL
mfr. no.   Cerilliant®
concentration   1.0 mg/mL in methanol
format   single component solution
storage temp.   −20°C
SMILES string   O=C1CC[C@@]2([H])[C@]3([H])CC[C@@]4([H])C([2H])([2H])[C@H](O[C@@H]5O[C@H](C(O)=O)[C@@H](O)[C@H](O)[C@H]5O)C([2H])([2H])C[C@]4(C)[C@@]3([H])CC[C@@]21C
InChI   1S/C25H38O8/c1-24-9-7-13(32-23-20(29)18(27)19(28)21(33-23)22(30)31)11-12(24)3-4-14-15-5-6-17(26)25(15,2)10-8-16(14)24/h12-16,18-21,23,27-29H,3-11H2,1-2H3,(H,30,31)/t12-,13+,14-,15-,16-,18-,19-,20+,21-,23+,24-,25-/m0/s1/i7D2,11D2
InChI key   VFUIRAVTUVCQTF-RDSFDFKRSA-N

Description

General description

An internal standard suitable for LC/MS or GC/MS applications with androsterone and androsterone glucuronide such as diagnostic testing for diagnosis and treatment of gonadal and adrenal diseases or sports testing to monitor testosterone doping. This Certified Spiking Solution® provides added value in allowing for quantitation of androsterone glucuronide levels by LC-MS/MS without the need for hydrolysis.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Eyes
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
WGK 2
Flash Point(F) 
49.5 °F - closed cup
Flash Point(C) 
9.7 °C - closed cup
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

D-063 Dehydroepiandrosterone (DHEA) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
D-064 Dehydroepiandrosterone-D5 (DHEA-D5) (2,2,3,4,4-D5) solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
D-065 Dehydroepiandrosterone 3-sulfate (DHEAS) sodium salt solution, 1.0 mg/mL in methanol (as free sulfate), ampule of 1 mL, certified reference material, Cerilliant®
D-066 Dehydroepiandrosterone-D5-3-sulfate (DHEAS-D5) (2,2,3,4,4,-D5) sodium salt solution, 100 μg/mL in methanol (as free sulfate), ampule of 1 mL, certified reference material, Cerilliant®

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?