• USA Home
  • C-145 - Chenodeoxycholic acid

EMAIL THIS PAGE TO A FRIEND
C-145 Supelco

Chenodeoxycholic acid

500 μg/mL in methanol, certified reference material, Cerilliant®

Synonym: 3α,7α-Dihydroxy-5β-cholanic acid, 5β-Cholanic acid-3α,7α-diol, Chenodiol

  • CAS Number 474-25-9

  • Empirical Formula (Hill Notation) C24H40O4

  • Molecular Weight 392.57

  •  Beilstein/REAXYS Number 3219887

  •  EC Number 200-659-6

  •  MDL number MFCD00064142

  •  PubChem Substance ID 329774884

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Bile Acid Standards, Bile acids, Biomarkers and Metabolomics Standards,
Quality Level   300
grade   certified reference material
feature   Snap-N-Spike®/Snap-N-Shoot®
packaging   ampule of 1 mL
mfr. no.   Cerilliant®
concentration   500 μg/mL in methanol
mp   165-167 °C (lit.)
format   single component solution
storage temp.   −20°C
SMILES string   C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
InChI   1S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16-,17-,18+,19+,20-,22+,23+,24-/m1/s1
InChI key   RUDATBOHQWOJDD-BSWAIDMHSA-N

Description

General description

Chenodeoxycholic acid (CDCA) is a primary bile acid that facilitates the intestinal uptake of lipids and fat-soluble vitamins. Levels of chenodeoxycholic acid can serve as markers for inborn and acquired hepatobiliary disorders such as biliary atresia and familial intrahepatic cholestasis as well as inborn errors of bile acid synthesis and sclerosing cholangitis. This Certified Spiking Solution® is suitable as a starting material in preparation of linearity standards, calibrators, and controls for use in LC-MS/MS and GC/MS bile acid testing methods.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
Protocols & Articles

Articles

HPLC Analysis of Bile Acids on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Bile Acids on Ascentis® Express C18
Keywords: Chromatography, Clinical, High performance liquid chromatography

Related Products

related product

Product #

Description

Add to Cart

B-903 Bromazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
P-080 Phenazepam solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
P-093 Phenazepam-D4 solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?