• USA Home
  • D-100 - (±)-2,5-Dimethoxy-4-Iodoamphetamine hydrochloride solution

EMAIL THIS PAGE TO A FRIEND
D-100 Supelco

(±)-2,5-Dimethoxy-4-Iodoamphetamine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Purchase

Properties

Related Categories Amphetamines / Stimulants Standards, Analytical Standards, Analytical/Chromatography, Pharmaceuticals, Illicit Drugs & Alcohol
grade   certified reference material
InChI Key   QVFDMWGKHUFODK-UHFFFAOYSA-N
feature   (Snap-N-Spike®)
packaging   ampule of 1 mL
mfr. no.   Cerilliant®
drug control   Narcotic Licence Schedule E (Switzerland)
concentration   1.0 mg/mL in methanol (as free base)
Featured Industry   Pharmaceutical (small molecule)
format   single component solution
storage temp.   −20°C

Description

General description

A Certified Spiking Solution® suited for GC/MS or LC/MS phenethylamine testing applications in forensic analysis, clinical toxicology, or urine drug testing. DOI is a D-substituted amphetamine in the phenethylamine family of hallucinogenic drugs. This designer drug is an extremely potent psychedelic with a long duration of action.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation

Snap-N-Spike is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

Amphetamines-weight loss drugs

Sigma-Aldrich's Cerilliant brand provides the widest selection of Certified Spiking Solutions® for amphetamines and weight loss drugs including native and stable-labeled analogs of amphetamine, metha...
Keywords: Ion-exchange chromatography

Emerging Drugs of Abuse

The illicit drug market is dynamic with new drugs of abuse constantly appearing. In addition to synthetic cannabinoids and synthetic cathinones, recent emerging drugs of abuse include new analogs of ...
Keywords: Addition reactions

LC/MS (TOF) Analysis of 2C Amines Drugs of Abuse in Urine on Ascentis® Express HILIC after Solid Phase Extraction (SPE) using Supel™-Select SCX

From our library of Articles, Sigma-Aldrich presents LC/MS (TOF) Analysis of 2C Amines Drugs of Abuse in Urine on Ascentis® Express HILIC after Solid Phase Extraction (SPE) using Supel™-Select SCX
Keywords: Chromatography, Clinical, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Pharmaceutical, Sample preparations, Solid phase extractions

LC/MS Analysis of 2C Amines Drugs of Abuse on Ascentis® Express HILIC

From our library of Articles, Sigma-Aldrich presents LC/MS Analysis of 2C Amines Drugs of Abuse on Ascentis® Express HILIC
Keywords: Chromatography, Clinical, High performance liquid chromatography, Mass spectrometry

Medication Monitoring Testing

Prescription monitoring requires accurate reference standards to ensure consistent and reliable results for drug confirmation and monitoring compliance.
Keywords: Anti-depressants, Anticonvulsants, Cardiovascular, Clinical, Controlled substances, Diabetes, Forensic, Gas chromatography, High performance liquid chromatography, Ion-exchange chromatography, Mass spectrometry, Metabolites, Pharmaceutical, Schizophrenia

Novel Psychoactive Phenethylamines

Phenethylamines represent a class of compounds with documented psychedelic and stimulant effects. The 2C family, which includes structural analogs such as 2C-B and 2C-I, has been well known since the...
Keywords: Clinical, Forensic

Related Products

related product

Product #

Description

Add to Cart

B-026 4-Bromo-2,5-dimethoxyphenethylamine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
T-084 2,5-Dimethoxy-4-(n)-propylthiophenethylamine hydrochloride solution, 1.0 mg/mL in acetonitrile: water (9:1) (as free base), ampule of 1 mL, certified reference material, Cerilliant®
D-095 2,5-Dimethoxy-4-methylphenethylamine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
E-083 2,5-Dimethoxy-4-ethylphenylethylamine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
43917 SnapIT Ampule Opener, large, for 5-10ml, 10-15ml, 20-25ml ampoules
06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?