EMAIL THIS PAGE TO A FRIEND
D-126 Supelco

Deoxycholic acid

500 μg/mL in methanol, certified reference material, Cerilliant®

Synonym: 3α,12α-Dihydroxy-5β-cholanic acid, 7-Deoxycholic acid, Desoxycholic acid

  • CAS Number 83-44-3

  • Empirical Formula (Hill Notation) C24H40O4

  • Molecular Weight 392.57

  •  Beilstein/REAXYS Number 3219882

  •  EC Number 200-659-6

  •  MDL number MFCD00003673

  •  PubChem Substance ID 329798683

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Bile Acid Standards, Bile acids, Biomarkers and Metabolomics Standards,
Quality Level   300
grade   certified reference material
feature   Snap-N-Spike®/Snap-N-Shoot®
packaging   ampule of 1 mL
mfr. no.   Cerilliant®
concentration   500 μg/mL in methanol
mp   171-174 °C (lit.)
format   single component solution
storage temp.   −20°C
SMILES string   C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
InChI   1S/C24H40O4/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3/h14-21,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15-,16-,17+,18-,19+,20+,21+,23+,24-/m1/s1
InChI key   KXGVEGMKQFWNSR-LLQZFEROSA-N

Description

General description

This [TM="Certified Spiking Solution" is suitable for use as starting material in the preparation of linearity standards, calibrators, and controls in LC-MS/MS and GC/MS bile acid testing methods. Deoxycholic acid (DCA), also known as deoxycholate and cholanoic acid is a secondary bile acid that aids in the absorption of fats in the intestine. Mass spectrometry-based analysis of DCA is routinely performed in clinical diagnostic testing applications including neonatal testing of inborn errors of bile acid synthesis and differentiating among types of familial intrahepatic cholestasis.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
Protocols & Articles

Articles

HPLC Analysis of Bile Acids on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Bile Acids on Ascentis® Express C18
Keywords: Chromatography, Clinical, High performance liquid chromatography

Related Products

related product

Product #

Description

Add to Cart

C-146 Cholic acid, 500 μg/mL in methanol, certified reference material, Cerilliant®
C-145 Chenodeoxycholic acid, 500 μg/mL in methanol, certified reference material, Cerilliant®
U-002 Ursodeoxycholic acid-D4, 100 μg/mL in methanol, certified reference material, Cerilliant®
C-147 Chenodeoxycholic acid-D4, 100 μg/mL in methanol, certified reference material, Cerilliant®
C-148 Cholic acid-D4, 100 μg/mL in methanol, certified reference material, Cerilliant®
D-127 Deoxycholic acid-D4, 100 μg/mL in methanol, certified reference material, Cerilliant®

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?