EMAIL THIS PAGE TO A FRIEND
D-127 Supelco

Deoxycholic acid-D4

100 μg/mL in methanol, certified reference material, Cerilliant®

  • CAS Number 112076-61-6

  • Empirical Formula (Hill Notation) C24H36D4O4

  • Molecular Weight 396.60

  •  MDL number MFCD00209707

  •  PubChem Substance ID 329798684

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Bile Acid Standards, Bile acids, Biomarkers and Metabolomics Standards,
Quality Level   300
grade   certified reference material
feature   Snap-N-Spike®/Snap-N-Shoot®
packaging   ampule of 1 mL
mfr. no.   Cerilliant®
concentration   100 μg/mL in methanol
format   single component solution
storage temp.   −20°C
SMILES string   C[C@H](CCC(O)=O)[C@@]1([H])CC[C@@]2([H])[C@]3([H])CC[C@]4([H])C([2H])([2H])[C@H](O)C([2H])([2H])C[C@]4(C)[C@@]3([H])C[C@H](O)[C@@]21C
InChI   1S/C24H40O4/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3/h14-21,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15-,16-,17+,18-,19+,20+,21+,23+,24-/m1/s1/i10D2,12D2
InChI key   KXGVEGMKQFWNSR-FCSCGBJGSA-N

Description

General description

A Certified Spiking Solution® suitable as an internal standard for quantitation of deoxycholic acid (DCA) levels in serum or other biological matrices by LC/MS or GC/MS for pharmaceutical research and clinical diagnostic testing applications. Deoxycholic acid (DCA), also known as deoxycholate and cholanoic acid, is a secondary bile acid that aids in the absorption of fats in the intestine. Mass spectrometry-based analysis of DCA is routinely performed in clinical diagnostic testing applications including neonatal testing of inborn errors of bile acid synthesis and differentiating among types of familial intrahepatic cholestasis.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

U-001 Ursodeoxycholic acid, 500 μg/mL in methanol, certified reference material, Cerilliant®
D-126 Deoxycholic acid, 500 μg/mL in methanol, certified reference material, Cerilliant®
C-146 Cholic acid, 500 μg/mL in methanol, certified reference material, Cerilliant®
C-145 Chenodeoxycholic acid, 500 μg/mL in methanol, certified reference material, Cerilliant®
L-042 Lithocholic acid, 50 μg/mL in methanol, certified reference material, Cerilliant®
U-002 Ursodeoxycholic acid-D4, 100 μg/mL in methanol, certified reference material, Cerilliant®
C-147 Chenodeoxycholic acid-D4, 100 μg/mL in methanol, certified reference material, Cerilliant®
C-148 Cholic acid-D4, 100 μg/mL in methanol, certified reference material, Cerilliant®
T-110 Taurocholic acid sodium salt, 500 μg/mL in methanol (as free sulfate), certified reference material, Cerilliant®

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?