• USA Home
  • D-146 - 5α-Dihydrotestosterone-D3 (16,16,17-D3) solution

EMAIL THIS PAGE TO A FRIEND
D-146 Supelco

5α-Dihydrotestosterone-D3 (16,16,17-D3) solution

10 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Synonym: 5α-DHT-D3 (16, 16, 17-D3) solution

  • CAS Number 79037-34-6

  • Empirical Formula (Hill Notation) C19H27O2D3

  • Molecular Weight 293.46

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Pharmaceuticals, Illicit Drugs & Alcohol, Steroids / Hormones
Quality Level   100
grade   certified reference material
feature   Snap-N-Spike®/Snap-N-Shoot®
packaging   ampule of 1 mL
mfr. no.   Cerilliant®
concentration   10 μg/mL in methanol
format   single component solution
storage temp.   −20°C
SMILES string   [2H]C1([2H])C[C@H]2[C@@H]3CCC4CC(=O)CC[C@]4(C)[C@H]3CC[C@]2(C)[C@@]1([2H])O
InChI   1S/C19H30O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12,14-17,21H,3-11H2,1-2H3/t12?,14-,15-,16-,17-,18-,19-/m0/s1/i6D2,17D
InChI key   NVKAWKQGWWIWPM-JUNPRZFISA-N

Description

General description

A stable-labeled internal standard of 5α-Dihydrotestosterone or DHT. DHT is an androgen or male sex hormone that has been implicated in a broad range of disease from male pattern baldness to benign prostatic hyperplasia and prostate cancer. As an anabolic steroid, DHT has also been used as a performance-enhancing drug to promote muscle growth.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

A-075 Androstenedione solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
D-073 5α-Dihydrotestosterone (DHT) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
E-058 Epitestosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
H-059 6β-Hydroxytestosterone solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
M-906 17α-Methyltestosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®
N-050 Nandrolone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
T-034 6β-Hydroxytestosterone-D3 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
T-037 Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
T-046 Testosterone-d3 (16, 16, 17-d3) solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
T-070 Testosterone-2,3,4-13C3 solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?