• USA Home
  • E-083 - 2,5-Dimethoxy-4-ethylphenylethylamine hydrochloride solution

EMAIL THIS PAGE TO A FRIEND
E-083 Supelco

2,5-Dimethoxy-4-ethylphenylethylamine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

  • CAS Number 923013-67-6

  • Empirical Formula (Hill Notation) C12H19NO2 · HCl

  • Molecular Weight 245.75

  •  EC Number 200-659-6

  •  NACRES NA.24

Purchase

Properties

Related Categories Amphetamines, Analytical Standards, Analytical/Chromatography, Pharmaceuticals, Illicit Drugs & Alcohol, Phenethylamines More...
Quality Level   100
grade   certified reference material
feature   (Snap-N-Spike®)
packaging   ampule of 1 mL
mfr. no.   Cerilliant®
drug control   Narcotic Licence Schedule E (Switzerland)
concentration   1.0 mg/mL in methanol (as free base)
Featured Industry   Pharmaceutical (small molecule)
format   single component solution
storage temp.   −20°C

Description

General description

2C-E, or 2,5-Dimethoxy-4-ethylphenethylamine, is a psychedelic drug and phenethylamine of the 2C series. Sold under street names including 2C-E or Eternity, the designer drug produces vivid visual hallucinations similar to those of LSD. This certified solution standard is suitable as a starting material for calibrators or controls in phenethylamine testing methods by GC/MS or LC/MS for applications in clinical toxicology,forensic analysis, or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

Snap-N-Spike is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Eyes
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
WGK 1
Flash Point(F) 
49.5 °F
Flash Point(C) 
9.7 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Amphetamines-weight loss drugs

Sigma-Aldrich's Cerilliant brand provides the widest selection of Certified Spiking Solutions® for amphetamines and weight loss drugs including native and stable-labeled analogs of amphetamine, metha...
Keywords: Ion-exchange chromatography

Emerging Drugs of Abuse

The illicit drug market is dynamic with new drugs of abuse constantly appearing. In addition to synthetic cannabinoids and synthetic cathinones, recent emerging drugs of abuse include new analogs of ...
Keywords: Addition reactions

Medication Monitoring Testing

Prescription monitoring requires accurate reference standards to ensure consistent and reliable results for drug confirmation and monitoring compliance.
Keywords: Anti-depressants, Anticonvulsants, Cardiovascular, Clinical, Controlled substances, Diabetes, Forensic, Gas chromatography, High performance liquid chromatography, Ion-exchange chromatography, Mass spectrometry, Metabolites, Pharmaceutical, Schizophrenia

Novel Psychoactive Phenethylamines

Phenethylamines represent a class of compounds with documented psychedelic and stimulant effects. The 2C family, which includes structural analogs such as 2C-B and 2C-I, has been well known since the...
Keywords: Clinical, Forensic

Related Products

related product

Product #

Description

Add to Cart

B-026 4-Bromo-2,5-dimethoxyphenethylamine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
B-049 (R)-(-)-Bromo-DragonFLY hydrochloride solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
C-122 2C-B-FLY hydrochloride solution, 1 mg/mL in acetonitrile: water (9:1), ampule of 1 mL, certified reference material, Cerilliant®
06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?