• USA Home
  • E-115 - Ethyl-β-D-glucuronide

EMAIL THIS PAGE TO A FRIEND
E-115 Supelco

Ethyl-β-D-glucuronide

10 mg/mL in methanol: water (1:1), certified reference material, ampule of 1 mL, Cerilliant®

  • CAS Number 17685-04-0

  • Empirical Formula (Hill Notation) C8H14O7

  • Molecular Weight 222.19

  •  MDL number MFCD09037574

Purchase

Properties

grade   certified reference material
InChI Key   IWJBVMJWSPZNJH-UQGZVRACSA-N
feature   Snap-N-Spike®/Snap-N-Shoot®
packaging   ampule of 1 mL
mfr. no.   Cerilliant®
concentration   10 mg/mL in methanol: water (1:1)
format   single component solution
storage temp.   −20°C

Description

General description

Ethyl-ß-D-glucuronide is a primary urinary metabolite of alcohol and can be used as a biomarker for the monitoring of alcohol consumption. This Snap-N-Spike® reference solution is applicable for use in LC/MS or GC/MS testing applications in clinical toxicology, forensic analysis, or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

Snap-N-Spike is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

RIDADR 
UN 1992 6.1(3) / PGIII

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

A-124 Multi-component Alcohol Mix, 250 μg/mL each component in H2O, certified reference material, Cerilliant®
A-125 Multi-component Alcohol Mix, 2000 μg/mL each component in H2O, certified reference material, Cerilliant®
A-127 Multi-component Alcohol Mix, in H2O (varied conc.), certified reference material, Cerilliant®
E-116 Ethyl sulfate sodium salt, 10 mg/mL in methanol: water (1:1), certified reference material, ampule of 1 mL, Cerilliant®
A-054 Multicomponent Alcohol Calibration Kit (C1-C3) solution, in H2O (varied conc.), ampule of 3 × 1.2 mL, certified reference material, Cerilliant®
A-056 Multicomponent Alcohol Mix 1000, 1000 μg/mL each component in H2O, ampule of 1.2 mL, certified reference material, Cerilliant®
A-057 Multicomponent Alcohol Mix 500, 500 μg/mL each component in H2O, ampule of 1.2 mL, certified reference material, Cerilliant®
A-061 Multicomponent Alcohol Mix-4000, 4000 μg/mL each component in H2O, ampule of 1.2 mL, certified reference material, Cerilliant®
A-076 Multi-Component Alcohol Mix-100, 100 μg/mL each component in H2O, ampule of 1.2 mL, certified reference material, Cerilliant®
E-015 Ethyl-β-D-glucuronide, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
E-016 Ethyl-β-D-glucuronide, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
E-048 Ethyl-β-D-glucuronide-(ethyl-d5), 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
E-063 Ethyl-β-D-glucuronide-(ethyl-d5), 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
E-064 Ethyl sulfate sodium salt, 1.0 mg/mL in methanol (as ethyl sulfate), ampule of 1 mL, certified reference material, Cerilliant®
E-066 Ethyl-d5 sulfate sodium salt, 1.0 mg/mL in methanol (as ethyl sulfate), ampule of 1 mL, certified reference material, Cerilliant®

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?