• USA Home
  • ERD-119 - O,O-Diethyl hydrogen thiophosphate potassium salt solution

EMAIL THIS PAGE TO A FRIEND
ERD-119 Supelco

O,O-Diethyl hydrogen thiophosphate potassium salt solution

1000 μg/mL in methanol (as free acid), ampule of 1.2 mL, certified reference material, Cerilliant®

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs), Certified Reference Materials from Cerilliant,
Quality Level   100
grade   certified reference material
InChI Key   OVJCAYMARFNMQB-UHFFFAOYSA-M
feature   Snap-N-Spike®/Snap-N-Shoot®
packaging   ampule of 1.2 mL
mfr. no.   Cerilliant®
concentration   1000 μg/mL in methanol (as free acid)
Featured Industry   Environmental
format   single component solution
storage temp.   −20°C

Description

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

Snap-N-Shoot is a registered trademark of Cerilliant Corporation

Snap-N-Spike is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Chemical Warfare Verification Standards

Sigma-Aldrich's Cerilliant brand offers the most comprehensive list of standards specifically designed for the analysis or detection of chemical weapons, Chemical Weapons Convention Treaty verificati...
Keywords: Ion-exchange chromatography, Metabolites

Pesticides

Sigma-Aldrich's Cerilliant brand offers an extensive line of pesticides certified reference standards for various applications. We supply single-component Snap-N-Shoot® solutions and reference materi...
Keywords: Ion-exchange chromatography, Metabolites, Pesticides

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?