• USA Home
  • ERD-148S - Dinitroethylene glycol solution

EMAIL THIS PAGE TO A FRIEND
ERD-148S Supelco

Dinitroethylene glycol solution

1000 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material, Cerilliant®

Synonym: Dinitroethylene glycol, EGDN

  • CAS Number 628-96-6

  • Empirical Formula (Hill Notation) C2H4N2O6

  • Molecular Weight 152.06

  •  MDL number MFCD01310543

  •  PubChem Substance ID 329799588

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs), Certified Reference Materials from Cerilliant,
grade   certified reference material
InChI Key   UQXKXGWGFRWILX-UHFFFAOYSA-N
feature   Snap-N-Spike®/Snap-N-Shoot®
packaging   ampule of 1.2 mL
mfr. no.   Cerilliant®
concentration   1000 μg/mL in acetonitrile
Featured Industry   Environmental
format   single component solution
storage temp.   −20°C

Description

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

Snap-N-Shoot is a registered trademark of Cerilliant Corporation

Snap-N-Spike is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Nitroglycerins/Explosives Standards

Sigma-Aldrich's Cerilliant maintains a U.S. ATFE license for the manufacture of explosive materials. According to US Department of Transportation (DOT) regulations, these products are normally classi...

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?