EMAIL THIS PAGE TO A FRIEND
F-032 Supelco

Flunitrazepam-13C6

100 μg/mL in methanol, certified reference material, Cerilliant®

  • CAS Number 1622-62-4

  • Empirical Formula (Hill Notation) C1013C6H12FN3O3

  • Molecular Weight 319.24

  •  MDL number MFCD00057906

  •  PubChem Substance ID 329799655

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Benzodiazepines, Pharmaceuticals, Illicit Drugs & Alcohol
Quality Level   100
grade   certified reference material
feature   Snap-N-Spike®/Snap-N-Shoot®
packaging   ampule of 1 mL
mfr. no.   Cerilliant®
drug control   psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)
concentration   100 μg/mL in methanol
format   single component solution
storage temp.   −20°C
InChI   1S/C16H12FN3O3/c1-19-14-7-6-10(20(22)23)8-12(14)16(18-9-15(19)21)11-4-2-3-5-13(11)17/h2-8H,9H2,1H3
InChI key   PPTYJKAXVCCBDU-UHFFFAOYSA-N
Gene Information   human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562), GABRD(2563), GABRE(2564), GABRG1(2565), GABRG2(2566), GABRG3(2567), GABRP(2568), GABRQ(55879)

Description

General description

This stable-labeled internal standard is suitable for quantitation of flunitrazepam levels by LC-MS/MS methods for clinical toxicology, forensic analysis, urine drug testing, pain prescription monitoring or isotope dilution applications. Flunitrazepam is an intermediate-acting benzodiazepine marketed under the trade name Rohypnol as a hypnotic, sedative, anticonvulsant and skeletal muscle relaxant. For these analyses, 13C-labeled internal standards can eliminate potential issues of isotopic exchange and minimize separation of later-eluting analytes from a higher substituted deuterium-labeled internal standard.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
Protocols & Articles

Articles

Benzodiazepine Standards

Benzodiazepines are a broad class of psychoactive drugs prescribed worldwide for treating numerous conditions from anxiety, seizures, and insomnia to muscle spasms, epilepsy, and alcohol withdrawal.1...
Keywords: Clinical, Forensic, Metabolites, Pharmaceutical

Medication Monitoring Testing

Prescription monitoring requires accurate reference standards to ensure consistent and reliable results for drug confirmation and monitoring compliance.
Keywords: Anti-depressants, Anticonvulsants, Cardiovascular, Clinical, Controlled substances, Diabetes, Forensic, Gas chromatography, High performance liquid chromatography, Ion-exchange chromatography, Mass spectrometry, Metabolites, Pharmaceutical, Schizophrenia

Related Products

related product

Product #

Description

Add to Cart

N-117 Nifoxipam solution, 1 mg/mL (90:10 Acetonitrile: DMSO (v/v)), certified reference material, ampule of 1 mL, Cerilliant®
N-118 Nifoxipam-D4 solution, 100 μg/mL (90:10 Acetonitrile: DMSO (v/v)), certified reference material, ampule of 1 mL, Cerilliant®
A-111 7-Aminoflunitrazepam-13C6 solution, 100 μg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®
H-140 3-Hydroxyflunitrazepam solution, 1 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®
H-141 3-Hydroxyflunitrazepam-13C6 solution, 100 μg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®
A-911 7-Aminoflunitrazepam solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
A-912 7-Aminoflunitrazepam solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
A-921 7-Aminoflunitrazepam-d7 solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
A-925 7-Aminoflunitrazepam-d7 solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
B-033 Benzodiazepine Multi-component Mixture-8 solution, 250 μg/mL each component in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
IMF-020 iMethod Test Kit for Benzodiazepines, kit of 22 × 1 mL ampules, certified reference material, Cerilliant®
D-918 N-Desmethylflunitrazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
D-925 N-Desmethylflunitrazepam-d4, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
F-907 Flunitrazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
F-915 Flunitrazepam-d7, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?