• USA Home
  • H-054 - 4-Hydroxytolbutamide solution

EMAIL THIS PAGE TO A FRIEND
H-054 Supelco

4-Hydroxytolbutamide solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

  • CAS Number 5719-85-7

  • Empirical Formula (Hill Notation) C12H18N2O4S

  • Molecular Weight 286.35

  •  EC Number 200-835-2

Purchase

Properties

Related Categories Additional Drugs, Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs),
grade   certified reference material
InChI Key   SJRHYONYKZIRPM-UHFFFAOYSA-N
feature   Snap-N-Spike®/Snap-N-Shoot®
packaging   ampule of 1 mL
mfr. no.   Cerilliant®
concentration   1.0 mg/mL in acetonitrile
Featured Industry   Pharmaceutical (small molecule)
format   single component solution
storage temp.   −20°C

Description

General description

4-Hydroxytolbutamide is a primary metabolite of tolbutamide, a potassium channel blocker used in the management of type-II diabetes. Tolbutamide is sold under the trade name Orinase. This certified Snap-N-Spike® solution is suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, urine drug testing, or pharmaceutical research including p450 testing.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

Snap-N-Shoot is a registered trademark of Cerilliant Corporation

Snap-N-Spike is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

Cytochrome P450 Standards

Sigma-Aldrich's Cerilliant brand provides a wide selection of Certified Solution Standards® for cytochrome P450 testing in pharmaceutical research and studies involving pharmacokinetics and drug meta...
Keywords: Ion-exchange chromatography, Metabolism, Metabolites, Pharmaceutical

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

43917 SnapIT Ampule Opener, large, for 5-10ml, 10-15ml, 20-25ml ampoules

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?