• USA Home
  • I-036 - Ibuprofen Impurity Mixture solution

EMAIL THIS PAGE TO A FRIEND
I-036 Supelco

Ibuprofen Impurity Mixture solution

25 μg/mL in acetonitrile (each component), certified reference material, ampule of 1 mL, Cerilliant®

  • Empirical Formula (Hill Notation) C11H14O2

  • Molecular Weight 178.23

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Ibuprofen Impurities, Pharmaceutical Impurities, Pharmacopeia & Metrological Institutes Standards More...
grade   certified reference material
feature   Snap-N-Spike®/Snap-N-Shoot®
packaging   ampule of 1 mL
mfr. no.   Cerilliant®
concentration   25 μg/mL in acetonitrile (each component)
Featured Industry   Pharmaceutical (small molecule)
format   multi-component solution
storage temp.   −20°C

Description

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

Analyte

Description

Product #

Add to Cart

4-ISOBUTYLACETOPHENONE(Ibuprofen Related Compound C)(Ibuprofen Impurity E), Pharmaceutical Secondary Standard; Certified Reference Material SDS PHR1146
Ibuprofen Related Compound J, Pharmaceutical Secondary Standard; Certified Reference Material SDS PHR1978
Ibuprofen Impurity A, Pharmaceutical Secondary Standard; Certified Reference Material SDS PHR1933
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material SDS PHR1004
Safety & Documentation

Safety Information

RIDADR 
UN 1648 3 / PGII

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

A-136 4-Aminophenol, 250 μg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®
A-134 Acetaminophen (Paracetamol) Impurity Mixture solution, 25 μg/mL in methanol (each component), certified reference material, ampule of 1 mL, Cerilliant®
A-135 4′-Acetoxyacetanilide (Acetaminophen RCA) solution, 250 μg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®
C-166 4-Chloroacetanilide (Acetaminophen RCJ) hydrochloride solution, 250 μg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®
M-195 Metformin Impurity Mixture hydrochloride solution, 50 μg/mL in methanol (each component), certified reference material, ampule of 1 mL, Cerilliant®
M-196 Cyanoguanidine (Metformin Related Compound A) solution, 50 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®
O-048 Omeprazole Impurity Mixture solution, 60 μg/mL in acetonitrile (each component), certified reference material, ampule of 1 mL, Cerilliant®
I-037 Ibuprofen Impurity B (PhEur) solution, 100 μg/mL in acetonitrile: water (9:1) (as free acid), certified reference material, ampule of 1 mL, Cerilliant®
P-140 Pramipexole Chiral System Suitability Solution solution, 10 μg/mL (85:15:0.1 Hexane:Ethanol:DEA), certified reference material, ampule of 1 mL, Cerilliant®
P-141 Pramipexole Organic Impurity System Suitability Mixture solution, certified reference material, ampule of 1 mL, Cerilliant®

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?