• USA Home
  • L-038 - Levofloxacin solution

EMAIL THIS PAGE TO A FRIEND
L-038 Supelco

Levofloxacin solution

1.0 mg/mL in acetonitrile, certified reference material, Cerilliant®

  • CAS Number 100986-85-4

  • Empirical Formula (Hill Notation) C18H20FN3O4

  • Molecular Weight 361.37

  •  MDL number MFCD00865049

  •  PubChem Substance ID 329817356

  •  NACRES NA.24

Purchase

Properties

Quality Level   100
  300
grade   certified reference material
feature   Snap-N-Spike®/Snap-N-Shoot®
packaging   ampule of 1 mL
mfr. no.   Cerilliant®
concentration   1.0 mg/mL in acetonitrile
format   single component solution
storage temp.   −20°C
InChI   1S/C18H20FN3O4/c1-10-9-26-17-14-11(16(23)12(18(24)25)8-22(10)14)7-13(19)15(17)21-5-3-20(2)4-6-21/h7-8,10H,3-6,9H2,1-2H3,(H,24,25)/t10-/m0/s1
InChI key   GSDSWSVVBLHKDQ-JTQLQIEISA-N

Description

Application

Levofloxacin is a broad-spectrum antibiotic that is effective in treating infections caused by gram-positive and gram-negative pathogens. This Certified Snap-N-Spike® solution standard is suitable as starting material for calibrators, controls, or linearity standards for therapeutic drug monitoring or clinical and diagnostic testing of levofloxacin in patient blood, serum, or plasma samples by LC-MS/MS, LC or GC/MS.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

DARCO is a registered trademark of Norit Americas, Inc.

Snap-N-Spike is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

RIDADR 
UN 1648 3 / PGII
Protocols & Articles

Articles

HPLC Analysis of Antibiotics (Fluoroquinolones from Tablets) on Discovery® HS C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Antibiotics (Fluoroquinolones from Tablets) on Discovery® HS C18
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Antibiotics on Discovery® C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Antibiotics on Discovery® C18
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?