• USA Home
  • M-176 - MDPPP hydrochloride solution

EMAIL THIS PAGE TO A FRIEND
M-176 Supelco

MDPPP hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym: 3′,4′-Methylenedioxy-α-pyrrolidinopropiophenone hydrochloride solution

  • CAS Number 24698-57-5

  • Empirical Formula (Hill Notation) C14H17NO3 · HCl

  • Molecular Weight 283.75

  •  NACRES NA.24

Purchase

Properties

Related Categories Amphetamines, Analytical Standards, Analytical/Chromatography, Cathinones, Pharmaceuticals, Illicit Drugs & Alcohol More...
Quality Level   100
grade   certified reference material
feature   (Snap-N-Spike®)
packaging   ampule of 1 mL
mfr. no.   Cerilliant®
drug control   Narcotic Licence Schedule E (Switzerland)
concentration   1.0 mg/mL in methanol (as free base)
Featured Industry   Pharmaceutical (small molecule)
storage temp.   −20°C

Description

General description

A Certified Spiking Solution® suitable for use as starting material in calibrators and controls for LC/MS or GC/MS MDPPP testing methods in clinical toxicology, forensic analysis, or urine drug testing applications. 3’,4’-Methylenedioxy-a-pyrrolidinopropiophenone, also known as MDPPP, is a synthetic cathinone found in numerous bath salt blends under the street name NRG-1. This designer drug first emerged in Germany in the late 1990s and early 2000s as an ingredient in imitation ecstasy pills.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation

Snap-N-Spike is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Eyes
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
WGK 1
Flash Point(F) 
49.5 °F - closed cup
Flash Point(C) 
9.7 °C - closed cup

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Amphetamines-weight loss drugs

Sigma-Aldrich's Cerilliant brand provides the widest selection of Certified Spiking Solutions® for amphetamines and weight loss drugs including native and stable-labeled analogs of amphetamine, metha...
Keywords: Ion-exchange chromatography

Emerging Drugs of Abuse

The illicit drug market is dynamic with new drugs of abuse constantly appearing. In addition to synthetic cannabinoids and synthetic cathinones, recent emerging drugs of abuse include new analogs of ...
Keywords: Addition reactions

HPLC Analysis of 3',4'-Methylenedioxy-α-pyrrolidinopropiophenone (MDPPP) Enantiomers on Astec® CHIROBIOTIC® V2

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of 3',4'-Methylenedioxy-α-pyrrolidinopropiophenone (MDPPP) Enantiomers on Astec® CHIROBIOTIC® V2
Keywords: Chromatography, High performance liquid chromatography

Medication Monitoring Testing

Prescription monitoring requires accurate reference standards to ensure consistent and reliable results for drug confirmation and monitoring compliance.
Keywords: Anti-depressants, Anticonvulsants, Cardiovascular, Clinical, Controlled substances, Diabetes, Forensic, Gas chromatography, High performance liquid chromatography, Ion-exchange chromatography, Mass spectrometry, Metabolites, Pharmaceutical, Schizophrenia

UHPLC Analysis of Flakka Compounds on Ascentis® Express HILIC

From our library of Articles, Sigma-Aldrich presents UHPLC Analysis of Flakka Compounds on Ascentis® Express HILIC
Keywords: Chromatography, High performance liquid chromatography, Separation, Solvents

UHPLC Analysis of Flakka and Related Compounds on Ascentis® Express RP-Amide in Reversed-Phase Mode

From our library of Articles, Sigma-Aldrich presents UHPLC Analysis of Flakka and Related Compounds on Ascentis® Express RP-Amide in Reversed-Phase Mode
Keywords: Chromatography, High performance liquid chromatography, Separation, Solvents

UHPLC Analysis of Flakka and Related Compounds on Titan™ Silica in HILIC Mode

From our library of Articles, Sigma-Aldrich presents UHPLC Analysis of Flakka and Related Compounds on Titan™ Silica in HILIC Mode
Keywords: Chromatography, High performance liquid chromatography, Separation, Solvents

“bath salt” Synthetic Cathinones Certified Spiking Solutions®

Cathinone-based stimulants emerged as a regulatory and societal threat in the European Union, Asia, and US in late 2010. Marketed as “bath salts” at head shops and on the internet, synthetic cathinon...
Keywords: Ion-exchange chromatography, Metabolites

Related Products

related product

Product #

Description

Add to Cart

C-172 4-Cl-alpha-PVP hydrochloride solution, 1 mg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant®
C-173 4-Cl-alpha-PPP hydrochloride solution, 1 mg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant®
T-121 TH-PVP hydrochloride solution, 1 mg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant®
M-146 3,4-Methylenedioxypyrovalerone HCl (MDPV) solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
M-150 3,4-Methylenedioxypyrovalerone-d8 HCl (MDPV-d8 HCl) solution, 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
M-154 MDPBP hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
N-067 Naphyrone hydrochloride, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
P-081 Pyrovalerone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
N-085 Naphyrone-D5 hydrochloride solution, 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
P-090 α-Pyrrolidinovalerophenone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
M-175 MDPBP-D8 hydrochloride solution, 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
P-101 α-Pyrrolidinovalerophenone-D8 hydrochloride solution, 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?