• USA Home
  • NMID607 - Etiocholanolone glucuronide

EMAIL THIS PAGE TO A FRIEND
NMID607

Etiocholanolone glucuronide

NMI Australia

  • CAS Number 3602-09-3

  • Empirical Formula (Hill Notation) C25H37O8Na

  • Molecular Weight 488.55

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Clinical Testing, Endogenous steroids and metabolites, NMI Australia,
Quality Level   100
grade   NMIA Certified Reference Material
mfr. no.   NMI Australia
format   neat
storage temp.   2-8°C
InChI   1S/C25H38O8/c1-24-9-7-13(32-23-20(29)18(27)19(28)21(33-23)22(30)31)11-12(24)3-4-14-15-5-6-17(26)25(15,2)10-8-16(14)24/h12-16,18-21,23,27-29H,3-11H2,1-2H3,(H,30,31)/t12-,13-,14+,15+,16+,18+,19+,20-,21+,23-,24+,25+/m1/s1
InChI key   VFUIRAVTUVCQTF-SDHZCXLISA-N
Safety & Documentation

Safety Information

WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Steroid Standards

Over the past 20 years the prevalence rate of illicit anabolic steroid use among US high school students has ranged from 2.2-6.1%.1 Anabolic steroids such as androstene, androsterone, boldenone and b...
Keywords: Cancer, Clinical, Clinical Chemistry, Diagnostic, Diseases, Endocrinology, High performance liquid chromatography, Hormones, Ion-exchange chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Metabolites

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?