EMAIL THIS PAGE TO A FRIEND
NMID826

Cocaine base

NMI Australia

  • CAS Number 50-36-2

  • Empirical Formula (Hill Notation) C17H21NO4

  • Molecular Weight 303.35

  •  EC Number 200-032-7

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Clinical Testing, Cocaine and related compounds, Cocaine, metabolites and related compounds,
Quality Level   100
grade   NMIA Certified Reference Material
mfr. no.   NMI Australia
format   neat
storage temp.   room temp

Description

General description

A reference material suitable for use in LC/MS or GC/MS testing applications for forensic analysis, urine drug testing, or clinical toxicology. Cocaine is a tropane alkaloid and illicit drug obtained from the leaves of the coca plant. This popular recreational drug is a powerful stimulant of the central nervous system with appetite suppressant and topical anesthetic properties.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Central nervous system
RIDADR 
UN 1544PSN1 6.1 / PGIII
WGK Germany 
WGK 3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?