EMAIL THIS PAGE TO A FRIEND
R-016 Supelco

Ramelteon solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs), Certified Reference Materials from Cerilliant,
grade   certified reference material
InChI Key   YLXDSYKOBKBWJQ-LBPRGKRZSA-N
feature   Snap-N-Spike®/Snap-N-Shoot®
packaging   ampule of 1 mL
mfr. no.   Cerilliant®
concentration   1.0 mg/mL in acetonitrile
Featured Industry   Pharmaceutical (small molecule)
format   single component solution
storage temp.   −20°C

Description

General description

A certified reference standard suitable as a starting material for preparation of calibrators and controls in GC/MS or LC/MS testing applications with pharmaceutical research, clinical toxicology, urine drug testing, or forensic analysis. Marketed and sold under the trade name Rozerem®, ramelteon is a newer sleep aid approved by the FDA for treatment of insomnia, particularly delayed sleep onset. The drug’s mode of action is unique for a sleep drug in that it does not bind to GABA receptors like the nonbenzodiazepines zolpidem, zopiclone, and zaleplon.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

Rozerem is a registered trademark of Takeda Pharmaceutical Company Limited

Snap-N-Shoot is a registered trademark of Cerilliant Corporation

Snap-N-Spike is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

43917 SnapIT Ampule Opener, large, for 5-10ml, 10-15ml, 20-25ml ampoules

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?