EMAIL THIS PAGE TO A FRIEND
SCB-014 Supelco

δ-BHC

vial of 1 g, analytical standard, Cerilliant®

Synonym: delta-BHC

  • CAS Number 319-86-8

  • Empirical Formula (Hill Notation) C6H6Cl6

  • Molecular Weight 290.83

  •  EC Number 206-272-9

  •  NACRES NA.24

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs), Certified Reference Materials from Cerilliant,
grade   analytical standard
form   neat
packaging   vial of 1 g
mfr. no.   Cerilliant®
Featured Industry   Agriculture
Environmental
Forensics and Toxicology
format   neat
storage temp.   room temp
SMILES string   Cl[C@H]1[C@@H](Cl)[C@@H](Cl)[C@H](Cl)[C@@H](Cl)[C@@H]1Cl
InChI   1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4+,5-,6-
InChI key   JLYXXMFPNIAWKQ-GPIVLXJGSA-N

Description

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Pesticides

Sigma-Aldrich's Cerilliant brand offers an extensive line of pesticides certified reference standards for various applications. We supply single-component Snap-N-Shoot® solutions and reference materi...
Keywords: Ion-exchange chromatography, Metabolites, Pesticides

The Definitive Guide to Cannabis Testing

The cannabinoids found in the Cannabis plant, commonly referred to as marijuana, have grown in popularity for treating a variety of ailments from arthritis, glaucoma and chronic pain to malnutrition,...
Keywords: Cancer, Chromatography, Clinical, Controlled substances, Diseases, Forensic, Gas chromatography, Genetic, Ion-exchange chromatography, Metabolites, Pesticides, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?