• USA Home
  • T-506 - Testosterone CRM in Serum solution

EMAIL THIS PAGE TO A FRIEND
T-506 Supelco

Testosterone CRM in Serum solution

52.5 ng/dL (Stripped Serum), certified reference material, ampule of 1 mL, Cerilliant®

  • Empirical Formula (Hill Notation) C19H28O2

  • Molecular Weight 288.42

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Certified Reference Materials in Matrix, Pharmaceuticals, Illicit Drugs & Alcohol
Quality Level   100
grade   certified reference material
packaging   ampule of 1 mL
mfr. no.   Cerilliant®
concentration   52.5 ng/dL (Stripped Serum)
format   single component solution
storage temp.   −20°C

Description

General description

Certified reference material of testosterone in stripped serum for clinical research applications by mass spectrometry. Individual levels of testosterone in stripped serum are offered separately at concentrations ranging from 2 to 2,000 ng/dL. Each individual level is ready to use as a linearity standard or for calibration verification in LC-MS/MS testosterone methods. Separate options for a blank and 13C- or deuterium labeled internal standards of testosterone are also available.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Certified Reference Material of Testosterone in Serum for Clinical Research Mass Spectrometry Applications

The critical importance of testosterone in the physiological development and function of males and females underscores its significant role in endocrine disorders and disease. Testosterone test resul...
Keywords: AGE, Cancer, Clinical, Clinical Chemistry, Diagnostic, Endocrinology, Forensic, High performance liquid chromatography, Immunoassay, Ion-exchange chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Pharmaceutical

Related Products

related product

Product #

Description

Add to Cart

T-500 Testosterone CRM in Serum solution, ampule of 1 mL (Blank), certified reference material, Cerilliant®
T-502 Testosterone CRM in Serum solution, 4 ng/dL (Stripped Serum), certified reference material, ampule of 1 mL, Cerilliant®
T-509 Testosterone CRM in Serum solution, 750 ng/dL (Stripped Serum), certified reference material, ampule of 1 mL, Cerilliant®
T-510 Testosterone CRM in Serum solution, 2000 ng/dL (Stripped Serum), certified reference material, ampule of 1 mL, Cerilliant®
D-073 5α-Dihydrotestosterone (DHT) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
D-077 5α-Dihydrotestosterone-D3 (16,16,17-D3) solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
E-058 Epitestosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
H-059 6β-Hydroxytestosterone solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
M-906 17α-Methyltestosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®
T-034 6β-Hydroxytestosterone-D3 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
T-037 Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
T-046 Testosterone-d3 (16, 16, 17-d3) solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
T-070 Testosterone-2,3,4-13C3 solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?