• USA Home
  • 05-550 - Anti-Cdc6 Antibody, clone DCS-180

EMAIL THIS PAGE TO A FRIEND
05-550 Sigma-Aldrich

Anti-Cdc6 Antibody, clone DCS-180

Detect Cdc6 with Anti-Cdc6 Antibody, clone DCS-180 (Mouse Monoclonal Antibody), that has been shown to work in IP & WB.

  •  eCl@ss 32160702

Purchase

Properties

Related Categories Antibodies, Primary Antibodies More...
packaging   100 µg (05-550)
gene symbol   CDC6(990)
trade name   Upstate
brand family   Upstate
format   Purified
molecular weight   62kDa
application(s)   Immunoprecipitation
  Western Blotting
isotype   IgG1
clone   Monoclonal Antibody
  clone DCS-180
host   Mouse
species reactivity   Human
  Mouse
NCBI accession no.   NM_001254
UniProt accession no.   Q99741
gene symbol   CDC18L
  HsCDC18
  HsCDC6
  cdc6
  p62
shipped in   2
Quality Assurance   routinely evaluated by immunoblot on HeLa nuclear extract or 3T3 nuclear extract

Description

Immunogen

Recombinant, full-length human cdc6

Specificity

cdc6

Physical form

10mM PBS, pH 7.4 and 0.05% sodium azide

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?