• USA Home
  • 06-118 - Anti-SHP-2/SHPTP-2 Antibody

EMAIL THIS PAGE TO A FRIEND
06-118 Sigma-Aldrich

Anti-SHP-2/SHPTP-2 Antibody

Detect SHP-2/SHPTP-2 using this Anti-SHP-2/SHPTP-2 Antibody validated for use in IC & WB.

Synonym: Tyrosine-protein phosphatase non-receptor type 11, SHP-2/SHPTP-2, Protein-tyrosine phosphatase SYP, SH-PTP2, SHP-2, Shp2

  •  eCl@ss 32160702

  •  NACRES NA.41

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, SG-SK
clone   polyclonal
biological source   rabbit
application(s)   immunocytochemistry: suitable
  western blot: suitable
species reactivity   mouse, human, human, mouse
species reactivity (predicted by homology)   rat (based on 100% sequence homology)
shipped in   dry ice
antibody product type   primary antibodies
purified by   affinity chromatography
mfr. no.   Upstate®
NCBI accession no.   NM_003179
UniProt accession no.   P35235
Gene Information   human ... PTPN11(5781)

Description

General description

Tyrosine-protein phosphatase non-receptor type 11 (EC 3.1.3.48; UniProt P35235; also known as Protein-tyrosine phosphatase SYP; SH-PTP2, SH2 domain-containing protein tyrosine phosphatase-2, SHP-2, Shp2) is encoded by the Ptpn11 (also known as Syp) gene (Gene ID 19247) in murine species. SHP-2/SHPTP-2 is a non-receptor protein tyrosine phosphatase involved in a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. SHP-2/SHPTP-2 contains two tandem Src homology-2 (SH2) domains that mediate substrates interaction, followed by a protein tyrosine phosphatase PTP domain. Peptide library-based study reveals similar substrate specificities of SHP-2 and the related SHP-1. Both enzymes exhibit preference for acidic and hydrophobic residues flanking the N- and C-terminal sides of target tyrosine residue. Mutations in PTPN11 gene are known to cause Noonan syndrome and acute myeloid leukemia in human.

Specificity

Recognizes SHP-2/SHPTP-2.

Immunogen

GST-fusion protein containing residues 2-216 of mouse SHP-2/SHPTP-2

Application

Immunocytochemistry Analysis: 4.0 µg/mL from a representative lot detected SHP-2/SHPTP-2 in HeLa, A431, HUVEC, and NIH/3T3 cells.

Research Category
Signaling

Research Sub Category
Cytoskeletal Signaling

Target description

64 kDa, apparent size increases to approximately 70 kDa upon tyrosine phosphorylation.

Physical form

Protein A Purfied

Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected SHP-2/SHPTP-2 in 10 µg of HeLa cell lysate.

Analysis Note

Control
Positive Control Included: Non-Stim A431 Cell Lysate (12-301)

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?