• USA Home
  • 07-227 - Anti-p34-Arc/ARPC2 Antibody

EMAIL THIS PAGE TO A FRIEND
07-227 Sigma-Aldrich

Anti-p34-Arc/ARPC2 Antibody

Anti-p34-Arc/ARPC2 Antibody detects level of p34-Arc/ARPC2 & has been published & validated for use in IC, IH & WB.

  •  eCl@ss 32160702

  •  NACRES NA.41

Purchase

Properties

Related Categories Antibodies, Primary Antibodies More...
clone   polyclonal
biological source   rabbit
application(s)   immunocytochemistry: suitable
  immunohistochemistry: suitable
  western blot: suitable
packaging   200 µg (07-2276-I)
NCBI accession no.   NM_005731
UniProt accession no.   O15144
mfr. no.   Upstate
Gene Information   human ... ARPC2(10109)
isotype   IgG
shipped in   dry ice

Description

Specificity

Recognizes p34-Arc/ARPC2 of the Arp2/3 complex.

Immunogen

Peptide corresponding to amino acids 285-298 of human Arc

Application

Research Category
Cell Structure

Research Sub Category
Cytoskeleton

Target description

34 kDa

Physical form

Format: Purified

Protein A Purfied

Protein A Purified immunoglobulin in 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide and 30% glycerol.

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Analysis Note

Control
Positive Control Included: Non-Stim A431 Cell Lysate (12-301)

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Quality

routinely evaluated by immunoblot on RIPA lysates from human A431, Jurkat, and mouse 3T3 cells

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?