• USA Home
  • 100110 - AAPH - CAS 2997-92-4 - Calbiochem

EMAIL THIS PAGE TO A FRIEND
100110 Millipore

AAPH - CAS 2997-92-4 - Calbiochem

A heat-sensitive, water-soluble, free radical initiator that induces oxidative stress by causing cellular damage associated with lipid peroxidation, protein modification, hemoglobin oxidation, and DNA strand breakage.

Synonym: AAPH - CAS 2997-92-4 - Calbiochem, ABAP, 2,2ʹ-Azobis-(2-amidinopropane), 2HCl, 2,2ʹ-Azobis-(2-methylpropionamidine), 2HCl

  • CAS Number 2997-92-4

  • Empirical Formula (Hill Notation) C8H18N6 · 2HCl

  • Molecular Weight 271.19

  •  MDL number MFCD00142725

Purchase

Properties

assay   ≥96% (titration)
form   solid
mfr. no.   Calbiochem®
storage condition   OK to freeze
  desiccated
  protect from light
storage conditions   -20C
color   white
solubility   water: 20 mg/mL
shipped in   ambient
InChI   1S/C8H18N6.ClH/c1-7(2,5(9)10)13-14-8(3,4)6(11)12;/h1-4H3,(H3,9,10)(H3,11,12);1H
InChI key   QMYCJCOPYOPWTI-UHFFFAOYSA-N

Description

Biochem/physiol Actions

Cell permeable: no

Product does not compete with ATP.

Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?