• USA Home
  • 1.05882 - Magnesium sulfate heptahydrate

EMAIL THIS PAGE TO A FRIEND
1.05882 SAFC

Magnesium sulfate heptahydrate

Emprove® Essential, Ph. Eur., BP, USP

Synonym: Epsom salts, bitter salt, epsom salt, sulfuric acid magnesium salt heptahydrate

  • CAS Number 10034-99-8

  • Linear Formula MgSO4 · 7H2O

  • Molecular Weight 246.47

  •  MDL number MFCD00149785

  •  EC Index Number 231-298-2

Purchase

Properties

Related Categories APIs (Active Pharmaceutical Ingredients) and Excipients, Biopharmaceutical Process Development and Manufacturing, Excipients, Liquid Dosage Forms, Mineral Salts,
Agency/Method   BP
  Ph. Eur.
  USP
Quality Level   500
vapor density   <0.01 (vs air)
vapor pressure   <0.1 mmHg ( 20 °C)
description   EMPROVE® ESSENTIAL Ph Eur,BP,USP
product line   Emprove® Essential
pH   5.0-8.0 (25 °C, 50 g/L in H2O)
solubility   710 g/L
density   1.68 g/cm3 at 20 °C
bulk density   900 kg/m3
SMILES string   O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
InChI   1S/Mg.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2
InChI key   WRUGWIBCXHJTDG-UHFFFAOYSA-L
storage conditions   Store at +5°C to +30°C.

Description

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
OM4508000

Emprove® Dossiers

Already an EMPROVE® Program customer? Sign in to access your EMPROVE® Dossiers

Sign In
What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Three document types are included with an Emprove® Program subscription:

  • Material Qualification Dossiers
  • Quality Management Dossiers
  • Operational Excellence Dossiers

Learn more about the benefits an Emprove® Program subscription can provide

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?