EMAIL THIS PAGE TO A FRIEND
1.06495 Supelco

Sodium hydroxide

Synonym: ‘Caustic soda’

Purchase

Properties

Related Categories Acids & Bases, Bases, Chemical Synthesis, Inorganic Bases, Synthetic Reagents More...
Quality Level   300
vapor density   >1 (vs air)
vapor pressure   <18 mmHg ( 20 °C)
  3 mmHg ( 37 °C)
mp   318 °C (lit.)
SMILES string   [OH-].[Na+]
InChI   1S/Na.H2O/h;1H2/q+1;/p-1
InChI key   HEMHJVSKTPXQMS-UHFFFAOYSA-M
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Articles

Analytical Method: Aluminium (total) in lime stone (Others)

Cat.No. 114944  Rectangular cells 50 mm Cat.No. 100594  Spectroquant® Aluminium Cell Test Cat.No. 114825  Spectroquant® Aluminium Test Cat.No. 100317  Hydrochloric acid 37 % for analysis Cat.No. 1064...
Keywords: Sample preparations

Analytical Method: Aluminium (total) in lime stone

Cat.No. 114947  Rectangular cells 20 mm and/or Cat.No. 114944  Rectangular cells 50 mm Cat.No. 100594  Spectroquant® Aluminium Cell Test Cat.No. 114825  Spectroquant® Aluminium Test Cat.No. 100317  H...
Keywords: Sample preparations

Analytical Method: Phosphorus (total) in iron oxide powder

Cat.No. 114944  Rectangular cells 50 mm Cat.No. 114543  Spectroquant® Phosphate Cell Test Cat.No. 114729  Spectroquant® Phosphate Cell Test Cat.No. 114848  Spectroquant® Phosphate Test Cat.No. 100716...
Keywords: Indicators, Melting, Sample preparations

Analytical Method: Silicon (total) in iron oxide powder

Cat.No. 114944  Rectangular cells 50 mm Cat.No. 114794  Spectroquant® Silicate (Silicic Acid) Test Cat.No. 100716  Sulfuric acid 25 % for analysis Cat.No. 106495  Sodium hydroxide for analysis Cat.No...
Keywords: Indicators, Melting, Sample preparations

Analytical Method: Silicon (total) in ores and ceramic materials

Cat.No. 114944  Rectangular cells 50 mm Cat.No. 114794  Spectroquant® Silicate (Silicic Acid) Test Cat.No. 101164  Ammonium fluoride for analysis Cat.No. 106495  Sodium hydroxide for analysis Cat.No....
Keywords: Sample preparations

Related Content

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?