• USA Home
  • 1.08808 - Zinc hydroxide carbonate

EMAIL THIS PAGE TO A FRIEND
1.08808 SAFC

Zinc hydroxide carbonate

pure (zinc carbonate basic)

Synonym: Zinc carbonate basic, Zinc hydroxide carbonate

Purchase

Properties

Related Categories APIs (Active Pharmaceutical Ingredients) and Excipients, Biopharmaceutical Process Development and Manufacturing, Excipients, Liquid Dosage Forms, Mineral Salts,
Quality Level   100
potency   >10000 mg/kg LD50, oral (Rat)
pH   9.5 (20 °C, 50 g/L in H2O, suspension)
mp   1970 °C
density   3.5 g/cm3 at 20 °C
bulk density   350‑450 kg/m3
storage conditions   Store at +5°C to +30°C.

Description

Analysis Note

Assay (complexometric, Zn): ≥ 58.0 %
Identity: passes test
Chloride (Cl): ≤ 0.05 %
Sulfate (SO₄): ≤ 0.5 %
As (Arsenic): ≤ 0.0005 %
Ca (Calcium): ≤ 0.5 %
Fe (Iron): ≤ 0.02 %
Pb (Lead): ≤ 0.002 %
Residual solvents (Ph Eur/USP): excluded by production process
Residue on ignition: 70.0 - 80.0 %

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?