EMAIL THIS PAGE TO A FRIEND
1.09327 Millipore

MAS 100 CG Ex

suitable for air monitoring

Purchase

Properties

Agency/Method   according to ISO 8573
feature   model (portable)
packaging   pkg of 1 kit
mfr. no.   MAS-100 CG Ex
application(s)   air monitoring: suitable (active microbial)
weight   10 kg
Featured Industry   Cosmetics
Food and Beverages
Pharmaceutical
storage conditions   Store at room temperature.

Description

General description

The MAS-100 CG Ex® has been specially developed for monitoring and measuring the microbiological contamination of compressed gases. The sampling method is based on the tried-and-tested Andersen impaction principle and accommodates the use of standard 90 to 100 mm Petri dishes. The compressed gases most commonly used in the industry are pre-programmed in the explosion proof device. The MAS-100 CG Ex® is the only instrument, which collects microorganisms under pressure and initiates automatic decompression after the collection cycle, thus preventing a sudden drop in pressure from damaging the microorganisms. The device is mobile, easy to use and powered via an integrated rechargeable battery. MAS-100 CG Ex® compressed gas microbial air sampler includes two sensors, one to measure mass flow and the other for pressure. The unit is pre-programmed for compressed air, nitrogen, carbon dioxide, argon and oxygen. Due to our unique mass flow sensor technology, after validation only a mathematical correction factor is required to sample other gases. The system measures the inflow of the gas and regulates it to a constant value of 100 liters/min and pressure built up ranging from 1.5 to 10 bar absolute. For low gas flow pipelines, a flow rate of 50 liters/min can be selected. The sampling head can be sterilized, and the instrument is an apparatus of category 3 according to EN 600079-14:1997. It can only be used in Zone 2 for gas groups IIA, IIB, IIC, comprising flammable substances which pose an explosion hazard in the temperature classes T1 to T6.

Application

The MAS-100 CG Ex® is suitable for microbial monitoring of compressed gasses, such as air, nitrogen, carbon dioxide, argon, oxygen and others. The microbial air sampling devices are extensively used in Pharmaceutical and Personal care industries as well as Food, Beverage and Environmental applications.

Components

MAS-100 CG Ex®, sampling head (100 L/min), hose, main charger, USB stick with all documentation and a travel case

Legal Information

MAS-100 CG Ex is a registered trademark of MBV AG

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Articles

Microbial Air Monitoring for the Pharma Industry: the MAS®-100 Series

The hygiene standard in a Pharmaceutical production environment is directly linked to the microbiological safety of the finished products. Make sure that your active microbial air sampling solutions ...
Keywords: Air sampling, Pharmaceutical, Sample preparations

Portable Microbial Air Samplers

The MAS-100 NT® and MAS-100 NT Ex® portable microbial air samplers are the industry standard for use in cleanrooms and critical environments. The MAS-100 NT® and MAS-100 NT Ex® systems have high coll...
Keywords: Air sampling, Filtration, Sample preparations

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?