• USA Home
  • 1.09525 - pH-indicator paper pH 1 - 10 Universal indicator

EMAIL THIS PAGE TO A FRIEND
1.09525 Supelco

pH-indicator paper pH 1 - 10 Universal indicator

pH-range 1 - 10

Purchase

Properties

Quality Level   100
pH-range   1 - 10
storage conditions   Store at +15°C to +25°C.

Description

General description

pH 1 - 2 - 3 - 4 - 5 - 6 - 7 - 8 - 9 - 10

pH indicator papers supplied in rolls are better suited for longterm storage, since they are protected against external influences. pH indicator papers are manufactured by impregnating high-quality filter paper with indicator or mixed-indicator solutions. Therefore, the paper is dryed, cut and packed into the corresponding sizes.

Analysis Note

Application test: conforms

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?