• USA Home
  • 1.15576 - LiChroCART Assembly tool

EMAIL THIS PAGE TO A FRIEND
1.15576 Supelco

LiChroCART Assembly tool

for use with LICHROCART® cartridge sealing ring assembly

Purchase

Properties

Related Categories Analytical/Chromatography, LPLC/TLC/Paper Chromatography, TLC Accessories, Thin Layer Chromatography (TLC)
Quality Level   100
application(s)   HPLC: suitable
compatibility   for use with LICHROCART® cartridge sealing ring assembly
storage conditions   no restrictions.

Description

General description

The package contains: 1 centering sleeve, 1 assembly tool, 1 tool for replacement of sealing rings from LiChroCART cartridges.

for replacement of sealing rings

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?