• USA Home
  • 1185-OP - OmniPur® Acrylamide Solution, 40% W/V - Calbiochem

EMAIL THIS PAGE TO A FRIEND
1185-OP EMD Millipore

OmniPur® Acrylamide Solution, 40% W/V - Calbiochem

Synonym: Propenamide

Purchase

Properties

Related Categories Biochemicals and Reagents More...
brand family   Calbiochem
packaging   500 ml in Plastic bottle (1185-500ML)
form   Clear, colorless liquid
shipped in   ambient
storage temp.   +15C to +30C
storage conditions   Ok to freeze

Description

Overview

Suitable nucleic acid and protein separation, standard resolution. Acrylamide in solution offers convenience and safety.

Specifications:
Acrylic acid: ≤0.005%
Conductivity: ≤5 µmho
DNase: None detected
pH at 25°C: 6.00-7.00
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?