• USA Home
  • 119101 - Anti-H+,K+-ATPase, α-Subunit (1019-1034) Rabbit pAb

EMAIL THIS PAGE TO A FRIEND
119101 Millipore

Anti-H+,K+-ATPase, α-Subunit (1019-1034) Rabbit pAb

Anti-H+,K+-ATPase, α-Subunit (1019-1034), rabbit polyclonal, recognizes the α-subunit of H+,K+-ATPase. It is validated for use in ELISA, WB, ICC, and IHC (paraffin sections).

Purchase

Properties

Related Categories Antibodies, Primary Antibodies More...
packaging   50 ul in Plastic ampoule (119101-50UL)
isotype   IgG
brand family   Calbiochem
form   Liquid
preservative   ≤0.1% sodium azide
clone   Polyclonal Antibody
host   Rabbit
species   canine, human, mouse, rat
species reactivity   Canine
  Human
  Mouse
  Rat
storage conditions   Avoid freeze/thaw
  Ok to freeze
shipped in   wet ice
storage conditions   = -70C

Description

Application

ELISA (see comments)

Immunoblotting (1:1000)

Immunocytochemistry (see comments)

Paraffin Sections (see comments)

General description

This product has been discontinued.



Recognizes the α-subunit of H+,K+-ATPase.

  • Antibody Target Gene Symbol: ATP4A
    • Target Synonym: ATP6A, Atpase, h+/k+ transporting, alpha polypeptide, Gastric H+/K+ ATPase Alpha, H+/K+ ATPASE, HKATPC, RATHKATPC
    • Entrez Gene Name: ATPase, H+/K+ exchanging, alpha polypeptide
    • Hu Entrez ID: 495 (Related Antibodies: 119102)
    • Mu Entrez ID: 11944
    • Rat Entrez ID: 24216
  • Rabbit polyclonal antibody supplied as undiluted serum. Recognizes the α-subunit of H+,K+-ATPase.

    Immunogen

    Porcine

    a synthetic peptide (GVRCCPGSWWDQGLYY) corresponding to amino acids 1019-1034 of porcine H+, K+-ATPase α-subunit

    Other Notes

    Exhibits some cross-reactivity with Na+, K+-ATPase at working dilution for H+,K+-ATPase. This antibody is also reported to work for ELISA, immunocytochemistry, and paraffin sections. Variables associated with assay conditions will dictate the proper working dilution.

    Smolka, A. 1992. Biophys. Biochem. Acta 1108, 75.

    Reconstitution

    Following initial thaw, aliquot and freeze (-70°C). Recommended dilution buffer is Tris buffered saline with 1% BSA, pH 7.5.

    Warning

    Standard Handling (A)

    Physical form

    Undiluted serum.

    Safety & Documentation

    Safety Information

    Safety Information for this product is unavailable at this time.

    Documents

    Certificate of Analysis

    ???PDP_SDS_LOT_NUMBER_ERROR_MSG???
    Protocols & Articles
    Related Products

    Technical Service:

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Bulk Ordering & Pricing:

    Need larger quantities for your development, manufacturing or research applications?