• USA Home
  • 122640 - ProteoExtract Albumin Removal Kit

EMAIL THIS PAGE TO A FRIEND
122640 Sigma-Aldrich

ProteoExtract Albumin Removal Kit

Purchase

Properties

usage   sufficient for 12 sample(s) (Plasma; 20-60 μl each)
mfr. no.   Calbiochem®
storage condition   OK to freeze
storage conditions   +2C to +8C
input   sample type serum
sample type plasma
sample type cerebrospinal fluid (CSF)
shipped in   ambient

Description

General description

Contains columns specifically designed for the rapid and efficient removal of >80% of albumin from serum body fluid samples. Removal of albumin allows for enhanced detection of low abundance proteins.

Packaging

1 kit in Glass bottle

Components

12 Disposable Columns packed with Albumin-Removal Resin, Binding Buffer, and a user protocol.

Warning

Toxicity: Standard Handling (A)

Other Notes

Hagman, C., et al. 2008. Anal. Chem.80, 1290.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?